Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACATTCCTACGCTGAGACTTAAT[A/G]TTTTCGTATACAGCTCTAGAAGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC8A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC8A - leucine rich repeat containing 8 family member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHYHD1 - phytanoyl-CoA dioxygenase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100876.1 | Intron | NP_001094346.1 | ||||
NM_001100877.1 | Intron | NP_001094347.1 | ||||
NM_174933.3 | Intron | NP_777593.2 |