Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGAAGGAGTTTGCTAGACACA[C/T]GGGGCACAAGGCATTCCAGACCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613036 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PMPCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PMPCA - peptidase, mitochondrial processing alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282944.1 | Intron | NP_001269873.1 | ||||
NM_001282946.1 | Intron | NP_001269875.1 | ||||
NM_015160.2 | Intron | NP_055975.1 | ||||
XM_005266059.4 | Intron | XP_005266116.1 | ||||
XM_011518417.2 | Intron | XP_011516719.1 | ||||
XM_017014543.1 | Intron | XP_016870032.1 |
SDCCAG3 - serologically defined colon cancer antigen 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |