Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGAAGCGAGCCCGGCAGATCAA[C/T]GTTTCCACAGATGACTCAGAGGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614533 MIM: 607795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC26 - cell division cycle 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRPF4 - pre-mRNA processing factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244926.1 | 410 | Silent Mutation | AAC,AAT | N,N 99 | NP_001231855.1 | |
NM_001322266.1 | 410 | UTR 5 | NP_001309195.1 | |||
NM_001322267.1 | 410 | UTR 5 | NP_001309196.1 | |||
NM_004697.4 | 410 | Silent Mutation | AAC,AAT | N,N 100 | NP_004688.2 |