Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTCCGCACCTACGTGATGCCCA[A/G]CGCACCTGTGGATGAGGCCATCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600440 | ||||||||||||||||||||
Literature Links: |
ENDOG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENDOG - endonuclease G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004435.2 | 1469 | Missense Mutation | AAC,AGC | N,S 251 | NP_004426.2 | |
XM_011518347.2 | 1469 | UTR 3 | XP_011516649.1 |
SPOUT1 - SPOUT domain containing methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016390.3 | 1469 | UTR 3 | NP_057474.2 | |||
XM_017014804.1 | 1469 | UTR 3 | XP_016870293.1 |