Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGATCCCCGTGTTCCTGTACAC[A/G]ACGTTTGGGCAGGTAAGGATCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616774 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIGA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIGA2 - mitoguardin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005252282.1 | 289 | Silent Mutation | ACA,ACG | T,T 28 | XP_005252339.1 | |
XM_011519117.1 | 289 | Silent Mutation | ACA,ACG | T,T 92 | XP_011517419.1 | |
XM_011519118.1 | 289 | UTR 5 | XP_011517420.1 |