Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCAGGAAAAAGAAGGTGTATGT[C/T]GGGGGTTTAGAGAGCAGGTATGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606443 MIM: 609471 MIM: 186745 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CREB3 - cAMP responsive element binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006368.4 | 978 | Silent Mutation | GTC,GTT | V,V 175 | NP_006359.3 |
GBA2 - glucosylceramidase beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6853 - microRNA 6853 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLN1 - talin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |