Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGAAACAGTATGATGTAGTGATT[C/G]GCATCACACTTCTCCAGCTCCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613774 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMSAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMSAP1 - calmodulin regulated spectrin associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015447.3 | 4053 | Silent Mutation | GCC,GCG | A,A 1509 | NP_056262.3 | |
XM_005263396.3 | 4053 | Silent Mutation | GCC,GCG | A,A 1520 | XP_005263453.1 | |
XM_005263397.1 | 4053 | Silent Mutation | GCC,GCG | A,A 1231 | XP_005263454.1 | |
XM_011518272.1 | 4053 | Silent Mutation | GCC,GCG | A,A 1376 | XP_011516574.1 | |
XM_011518273.2 | 4053 | Silent Mutation | GCC,GCG | A,A 1321 | XP_011516575.1 | |
XM_011518274.1 | 4053 | Silent Mutation | GCC,GCG | A,A 1310 | XP_011516576.1 | |
XM_017014301.1 | 4053 | Silent Mutation | GCC,GCG | A,A 1376 | XP_016869790.1 | |
XM_017014302.1 | 4053 | Silent Mutation | GCC,GCG | A,A 1376 | XP_016869791.1 |