Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGTCCCACGCTGCTCACAATGA[A/G]GCCCACAGCCTCAAATATGAACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602956 MIM: 614730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FANCG - Fanconi anemia complementation group G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGO - phosphatidylinositol glycan anchor biosynthesis class O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201484.1 | 2406 | Missense Mutation | CTC,TTC | L,F 638 | NP_001188413.1 | |
NM_032634.3 | 2406 | Missense Mutation | CTC,TTC | L,F 1055 | NP_116023.2 | |
NM_152850.3 | 2406 | Missense Mutation | CTC,TTC | L,F 638 | NP_690577.2 | |
XM_005251619.3 | 2406 | Missense Mutation | CTC,TTC | L,F 1055 | XP_005251676.1 | |
XM_017015222.1 | 2406 | Missense Mutation | CTC,TTC | L,F 1055 | XP_016870711.1 | |
XM_017015223.1 | 2406 | Missense Mutation | CTC,TTC | L,F 638 | XP_016870712.1 | |
XM_017015224.1 | 2406 | Missense Mutation | CTC,TTC | L,F 638 | XP_016870713.1 |