Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGATTGAGACCAATGAGCAGAAG[A/C]GTGGGTGAGGGTGGGGGCAGGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf24 - chromosome 9 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252195.1 | 646 | Intron | NP_001239124.1 | |||
NM_032596.3 | 646 | Intron | NP_115985.2 | |||
NM_147168.1 | 646 | Intron | NP_671697.1 | |||
NM_147169.2 | 646 | Intron | NP_671698.1 | |||
XM_005251616.4 | 646 | UTR 5 | XP_005251673.1 | |||
XM_005251618.4 | 646 | Intron | XP_005251675.1 | |||
XM_011518055.2 | 646 | Intron | XP_011516357.1 |
FAM219A - family with sequence similarity 219 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184940.1 | 646 | UTR 3 | NP_001171869.1 | |||
NM_001184941.1 | 646 | UTR 3 | NP_001171870.1 | |||
NM_001184942.1 | 646 | UTR 3 | NP_001171871.1 | |||
NM_001184943.1 | 646 | UTR 3 | NP_001171872.1 | |||
NM_001184945.1 | 646 | UTR 3 | NP_001171874.1 | |||
NM_147202.1 | 646 | UTR 3 | NP_671735.1 | |||
XM_005251402.4 | 646 | UTR 3 | XP_005251459.1 | |||
XM_011517797.2 | 646 | UTR 3 | XP_011516099.1 | |||
XM_011517798.2 | 646 | UTR 3 | XP_011516100.1 | |||
XM_011517799.2 | 646 | UTR 3 | XP_011516101.1 |