Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTACCACTTCTGCACCGTGTGG[A/C]CTTCCTTGATCACCTGTGTAAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C9orf131 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C9orf131 - chromosome 9 open reading frame 131 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040410.2 | 88 | Missense Mutation | GAC,GCC | D,A 51 | NP_001035500.1 | |
NM_001040411.2 | 88 | Missense Mutation | GAC,GCC | D,A 13 | NP_001035501.1 | |
NM_001040412.2 | 88 | Missense Mutation | GAC,GCC | D,A 38 | NP_001035502.1 | |
NM_001287391.1 | 88 | Missense Mutation | GAC,GCC | D,A 13 | NP_001274320.1 | |
NM_203299.3 | 88 | Missense Mutation | GAC,GCC | D,A 86 | NP_976044.2 |