Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATGATGAAGAGCATGGTGGCCA[C/G]CAGGGGCACCAGGGTGAGGTAGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613104 MIM: 606813 | ||||||||||||||||||||
Literature Links: |
CACFD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACFD1 - calcium channel flower domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC2A6 - solute carrier family 2 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145099.1 | 1265 | Intron | NP_001138571.1 | |||
NM_017585.3 | 1265 | Missense Mutation | CTG,GTG | L,V 400 | NP_060055.2 | |
XM_011518189.2 | 1265 | Missense Mutation | CTG,GTG | L,V 194 | XP_011516491.2 | |
XM_017014236.1 | 1265 | Intron | XP_016869725.1 | |||
XM_017014237.1 | 1265 | Missense Mutation | CTG,GTG | L,V 156 | XP_016869726.1 | |
XM_017014238.1 | 1265 | Missense Mutation | CTG,GTG | L,V 156 | XP_016869727.1 |