Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCACAACGCCTCCTACCTCACC[A/G]ACTTCCACAGCCAGGACGAGAGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604349 | ||||||||||||||||||||
Literature Links: |
LAMC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAMC3 - laminin subunit gamma 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006059.3 | 375 | Missense Mutation | AAC,GAC | N,D 93 | NP_006050.3 | |
XM_006716921.2 | 375 | Missense Mutation | AAC,GAC | N,D 93 | XP_006716984.1 | |
XM_011518121.1 | 375 | Missense Mutation | AAC,GAC | N,D 93 | XP_011516423.1 |