Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAACACACTGCTGGTCTCTTAC[A/C]GGGCCATTTCCTTGGTCTCCTGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC100506100 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100506100 - uncharacterized LOC100506100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZDHHC12 - zinc finger DHHC-type containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZER1 - zyg-11 related cell cycle regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006336.3 | 2483 | Missense Mutation | CGC,CTC | R,L 748 | NP_006327.2 | |
XM_005251645.2 | 2483 | Missense Mutation | CGC,CTC | R,L 748 | XP_005251702.1 | |
XM_005251646.2 | 2483 | Missense Mutation | CGC,CTC | R,L 746 | XP_005251703.1 | |
XM_011518122.2 | 2483 | Missense Mutation | CGC,CTC | R,L 748 | XP_011516424.1 | |
XM_017014185.1 | 2483 | Missense Mutation | CGC,CTC | R,L 748 | XP_016869674.1 | |
XM_017014186.1 | 2483 | Missense Mutation | CGC,CTC | R,L 746 | XP_016869675.1 | |
XM_017014187.1 | 2483 | Missense Mutation | CGC,CTC | R,L 746 | XP_016869676.1 | |
XM_017014188.1 | 2483 | Missense Mutation | CGC,CTC | R,L 746 | XP_016869677.1 | |
XM_017014189.1 | 2483 | Missense Mutation | CGC,CTC | R,L 685 | XP_016869678.1 | |
XM_017014190.1 | 2483 | Missense Mutation | CGC,CTC | R,L 685 | XP_016869679.1 |