Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATCAACATGGCTATGGTATTTA[A/G]TAATGGCCCAGCTTAGAGACTTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMOD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMOD1 - tropomodulin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166116.1 | 2204 | UTR 3 | NP_001159588.1 | |||
NM_003275.3 | 2204 | UTR 3 | NP_003266.1 |
TSTD2 - thiosulfate sulfurtransferase like domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139246.4 | 2204 | UTR 3 | NP_640339.4 | |||
XM_011518312.2 | 2204 | UTR 3 | XP_011516614.1 | |||
XM_017014342.1 | 2204 | UTR 3 | XP_016869831.1 |