Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCGCTGTTGCTGTACTCCTCGCC[A/C]CAGTCGATGGGGGCGCCCTCGGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614045 MIM: 610933 | ||||||||||||||||||||
Literature Links: |
FAM129B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM129B - family with sequence similarity 129 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035534.2 | 1798 | Missense Mutation | TGG,TGT | W,C 576 | NP_001030611.1 | |
NM_022833.3 | 1798 | Missense Mutation | TGG,TGT | W,C 589 | NP_073744.2 | |
XM_005252135.2 | 1798 | Missense Mutation | TGG,TGT | W,C 614 | XP_005252192.2 | |
XM_011518925.1 | 1798 | Missense Mutation | TGG,TGT | W,C 619 | XP_011517227.1 |
LRSAM1 - leucine rich repeat and sterile alpha motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |