Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAAGCTAGGAGGTGTCTCTTCAA[C/T]GGTATGAGGAAACAGTGCCATGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607699 | ||||||||||||||||||||
Literature Links: |
RNF20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF20 - ring finger protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019592.6 | 497 | Missense Mutation | ACG,ATG | T,M 43 | NP_062538.5 | |
XM_011518862.1 | 497 | Missense Mutation | ACG,ATG | T,M 43 | XP_011517164.1 |