Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCGGGCTGCAGGCGGCATCCTA[C/T]ACCTGGAGCTGCTGGTGGCCGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604134 MIM: 602930 | ||||||||||||||||||||
Literature Links: |
ADAMTS13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTS13 - ADAM metallopeptidase with thrombospondin type 1 motif 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139025.4 | 685 | Missense Mutation | CAC,TAC | H,Y 81 | NP_620594.1 | |
NM_139026.4 | 685 | Missense Mutation | CAC,TAC | H,Y 81 | NP_620595.1 | |
NM_139027.4 | 685 | Missense Mutation | CAC,TAC | H,Y 81 | NP_620596.2 | |
XM_011518176.2 | 685 | Intron | XP_011516478.1 | |||
XM_011518178.1 | 685 | Intron | XP_011516480.1 | |||
XM_011518179.1 | 685 | Intron | XP_011516481.1 | |||
XM_017014232.1 | 685 | Missense Mutation | CAC,TAC | H,Y 77 | XP_016869721.1 | |
XM_017014233.1 | 685 | UTR 5 | XP_016869722.1 | |||
XM_017014234.1 | 685 | Intron | XP_016869723.1 | |||
XM_017014235.1 | 685 | Missense Mutation | CAC,TAC | H,Y 81 | XP_016869724.1 |
REXO4 - REX4 homolog, 3'-5' exonuclease | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |