Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGATTGCTGCTAAAATTGATTCA[A/G]TTCCTCACTTGAATAATTCCACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603536 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUBP3 - far upstream element binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003934.1 | 220 | Missense Mutation | ATT,GTT | I,V 36 | NP_003925.1 | |
XM_005272232.1 | 220 | Missense Mutation | ATT,GTT | I,V 36 | XP_005272289.1 | |
XM_006717313.2 | 220 | UTR 5 | XP_006717376.1 | |||
XM_011519172.2 | 220 | Missense Mutation | ATT,GTT | I,V 36 | XP_011517474.1 | |
XM_011519173.2 | 220 | Intron | XP_011517475.1 | |||
XM_011519174.2 | 220 | UTR 5 | XP_011517476.1 | |||
XM_017015269.1 | 220 | UTR 5 | XP_016870758.1 | |||
XM_017015270.1 | 220 | UTR 5 | XP_016870759.1 | |||
XM_017015271.1 | 220 | UTR 5 | XP_016870760.1 | |||
XM_017015272.1 | 220 | Intron | XP_016870761.1 | |||
XM_017015273.1 | 220 | UTR 5 | XP_016870762.1 |