Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTTCAGGTTATTTAGGAGGATT[C/T]GATGGCAATTTTTTGTGGAACAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607423 | ||||||||||||||||||||
Literature Links: |
POMT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POMT1 - protein O-mannosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077365.1 | 448 | Silent Mutation | TTC,TTT | F,F 82 | NP_001070833.1 | |
NM_001077366.1 | 448 | Silent Mutation | TTC,TTT | F,F 28 | NP_001070834.1 | |
NM_001136113.1 | 448 | Silent Mutation | TTC,TTT | F,F 82 | NP_001129585.1 | |
NM_001136114.1 | 448 | UTR 5 | NP_001129586.1 | |||
NM_007171.3 | 448 | Silent Mutation | TTC,TTT | F,F 82 | NP_009102.3 | |
XM_005272156.1 | 448 | Silent Mutation | TTC,TTT | F,F 82 | XP_005272213.1 | |
XM_005272158.1 | 448 | Silent Mutation | TTC,TTT | F,F 28 | XP_005272215.1 | |
XM_005272159.1 | 448 | UTR 5 | XP_005272216.1 | |||
XM_005272162.2 | 448 | UTR 5 | XP_005272219.1 | |||
XM_006716932.1 | 448 | UTR 5 | XP_006716995.1 | |||
XM_011518140.1 | 448 | Silent Mutation | TTC,TTT | F,F 82 | XP_011516442.1 | |
XM_011518141.1 | 448 | Silent Mutation | TTC,TTT | F,F 82 | XP_011516443.1 | |
XM_011518142.1 | 448 | Silent Mutation | TTC,TTT | F,F 28 | XP_011516444.1 | |
XM_011518143.1 | 448 | Nonsense Mutation | CGA,TGA | R,* 47 | XP_011516445.1 | |
XM_011518145.1 | 448 | UTR 5 | XP_011516447.1 | |||
XM_017014203.1 | 448 | Silent Mutation | TTC,TTT | F,F 28 | XP_016869692.1 | |
XM_017014204.1 | 448 | UTR 5 | XP_016869693.1 | |||
XM_017014205.1 | 448 | UTR 5 | XP_016869694.1 |
PRRC2B - proline rich coiled-coil 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |