Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCACTCACCTGGTCCCCTTCTT[G/T]TTGGGCACTGTGTATGGGCGGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604884 MIM: 604030 | ||||||||||||||||||||
Literature Links: |
LOC100129034 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100129034 - uncharacterized LOC100129034 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK6 - NIMA related kinase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMB7 - proteasome subunit beta 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002799.3 | 756 | Missense Mutation | AAA,AAC | K,N 236 | NP_002790.1 |