Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCTCCTCCTCTTGGGAAAAGTT[C/T]AGCAACACTGACACAGCTTTTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602502 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC5L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARPC5L - actin related protein 2/3 complex subunit 5 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GOLGA1 - golgin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002077.3 | 2481 | Silent Mutation | CTA,CTG | L,L 724 | NP_002068.1 | |
XM_005251929.3 | 2481 | Silent Mutation | CTA,CTG | L,L 724 | XP_005251986.1 | |
XM_006717062.3 | 2481 | Silent Mutation | CTA,CTG | L,L 724 | XP_006717125.1 | |
XM_006717063.3 | 2481 | Silent Mutation | CTA,CTG | L,L 724 | XP_006717126.1 |