Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACCTCCGCCTTCCCACTAGTCC[C/T]GCAACCAGAAGAGCTGGACCAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616748 MIM: 611255 | ||||||||||||||||||||
Literature Links: |
ENTPD8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENTPD8 - ectonucleoside triphosphate diphosphohydrolase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033113.1 | 1326 | Missense Mutation | CAG,CGG | Q,R 494 | NP_001028285.1 | |
NM_198585.2 | 1326 | Missense Mutation | CAG,CGG | Q,R 457 | NP_940987.2 | |
XM_011518665.2 | 1326 | Missense Mutation | CAG,CGG | Q,R 602 | XP_011516967.1 | |
XM_011518666.2 | 1326 | Missense Mutation | CAG,CGG | Q,R 565 | XP_011516968.1 | |
XM_011518668.1 | 1326 | Missense Mutation | CAG,CGG | Q,R 367 | XP_011516970.1 |
NOXA1 - NADPH oxidase activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256067.1 | 1326 | Intron | NP_001242996.1 | |||
NM_001256068.1 | 1326 | Intron | NP_001242997.1 | |||
NM_006647.1 | 1326 | Intron | NP_006638.1 | |||
XM_011518158.2 | 1326 | Intron | XP_011516460.1 | |||
XM_011518159.1 | 1326 | Intron | XP_011516461.1 | |||
XM_011518160.1 | 1326 | Intron | XP_011516462.1 | |||
XM_017014220.1 | 1326 | Intron | XP_016869709.1 |