Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGGAAAATATCCCTGCGGTCTG[C/T]GAGGGGCTTGGGGGAGCAGTCAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107987125 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107987125 - uncharacterized LOC107987125 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017015410.1 | 8190 | Intron | XP_016870899.1 |
TTLL11 - tubulin tyrosine ligase like 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139442.1 | 8190 | Intron | NP_001132914.1 | |||
NM_194252.2 | 8190 | Intron | NP_919228.2 | |||
XM_005251728.2 | 8190 | UTR 3 | XP_005251785.1 | |||
XM_005251729.4 | 8190 | UTR 3 | XP_005251786.1 | |||
XM_006716966.3 | 8190 | Intron | XP_006717029.1 | |||
XM_017014309.1 | 8190 | Intron | XP_016869798.1 |