Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCACTACATCCCCGTTGGCATT[A/G]TAATACCGGGTCACATTTAGGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613210 MIM: 611424 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPH7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPH7 - diphthamide biosynthesis 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZMYND19 - zinc finger MYND-type containing 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138462.2 | 672 | Silent Mutation | TAC,TAT | Y,Y 150 | NP_612471.1 | |
XM_005266052.4 | 672 | Silent Mutation | TAC,TAT | Y,Y 211 | XP_005266109.1 | |
XM_017014255.1 | 672 | Intron | XP_016869744.1 |