Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTGTGCCCGCAGCCCTGCAGCT[C/T]GCCGGTGCCCTCGACCTGCCCGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612879 | ||||||||||||||||||||
Literature Links: |
MAMDC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAMDC2 - MAM domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153267.4 | 724 | Silent Mutation | CTC,CTT | L,L 15 | NP_694999.3 | |
XM_011518471.1 | 724 | Silent Mutation | CTC,CTT | L,L 15 | XP_011516773.1 | |
XM_011518475.2 | 724 | Silent Mutation | CTC,CTT | L,L 15 | XP_011516777.1 | |
XM_017014587.1 | 724 | Silent Mutation | CTC,CTT | L,L 15 | XP_016870076.1 |