Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAATGTCAAGTGCAGGAAAGACA[A/G]TATCTATTAAAAAAAAAAAGTCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603873 | ||||||||||||||||||||
Literature Links: |
PLAA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLAA - phospholipase A2 activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031689.2 | 2185 | Missense Mutation | ACT,ATT | T,I 609 | NP_001026859.1 | |
NM_001321546.1 | 2185 | Missense Mutation | ACT,ATT | T,I 586 | NP_001308475.1 | |
XM_011518072.2 | 2185 | Intron | XP_011516374.1 | |||
XM_017015319.1 | 2185 | Intron | XP_016870808.1 |