Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGATAAGAATTTTTTGTTTTTTG[G/T]GTAATTAGCAATAATATTTTAGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf147 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf147 - chromosome 9 open reading frame 147 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA1958 - KIAA1958 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287036.1 | Intron | NP_001273965.1 | ||||
NM_001287038.1 | Intron | NP_001273967.1 | ||||
NM_133465.3 | Intron | NP_597722.1 | ||||
XM_011518311.2 | Intron | XP_011516613.1 |