Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTTGGTAGAAAGACGCAGGATG[C/T]GACAGTCTGGAAATGCCCCTTATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
WDR31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR31 - WD repeat domain 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006615.2 | 1060 | Missense Mutation | NP_001006616.1 | |||
NM_001012361.3 | 1060 | Missense Mutation | NP_001012361.1 | |||
NM_145241.4 | 1060 | Missense Mutation | NP_660284.1 | |||
XM_011518194.2 | 1060 | Missense Mutation | XP_011516496.1 |