Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTCATTCTTTATATATACTCAA[C/T]GGTAATTTGAATCGTTAGGAATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176877 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPN3 - protein tyrosine phosphatase, non-receptor type 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145368.1 | 4646 | UTR 3 | NP_001138840.1 | |||
NM_001145369.1 | 4646 | UTR 3 | NP_001138841.1 | |||
NM_001145370.1 | 4646 | UTR 3 | NP_001138842.1 | |||
NM_001145371.1 | 4646 | UTR 3 | NP_001138843.1 | |||
NM_001145372.1 | 4646 | UTR 3 | NP_001138844.1 | |||
NM_002829.3 | 4646 | UTR 3 | NP_002820.3 | |||
XM_006717197.3 | 4646 | UTR 3 | XP_006717260.1 | |||
XM_006717199.3 | 4646 | UTR 3 | XP_006717262.1 | |||
XM_006717201.3 | 4646 | UTR 3 | XP_006717264.1 | |||
XM_006717202.3 | 4646 | UTR 3 | XP_006717265.1 | |||
XM_006717203.3 | 4646 | UTR 3 | XP_006717266.1 | |||
XM_006717204.3 | 4646 | UTR 3 | XP_006717267.1 | |||
XM_011518888.2 | 4646 | UTR 3 | XP_011517190.1 | |||
XM_011518889.2 | 4646 | UTR 3 | XP_011517191.1 | |||
XM_017014955.1 | 4646 | UTR 3 | XP_016870444.1 | |||
XM_017014956.1 | 4646 | UTR 3 | XP_016870445.1 | |||
XM_017014957.1 | 4646 | UTR 3 | XP_016870446.1 |