Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATCCACCAGAATCGCCTGTCGA[C/G]TGTTACAGAAGATGAAGACCAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603421 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSANTD3-TMEFF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSANTD3-TMEFF1 - MSANTD3-TMEFF1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198812.1 | 124 | Intron | NP_001185741.1 |
MURC - muscle related coiled-coil protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018116.2 | 124 | Missense Mutation | ACT,AGT | T,S 20 | NP_001018126.1 |
TMEFF1 - transmembrane protein with EGF like and two follistatin like domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003692.4 | 124 | Intron | NP_003683.2 |