Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGCCGGCTTTCCTTGGGTGCGC[G/T]CGCAGCTTTCTGTGCGCCAGTTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602580 MIM: 616528 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLGA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GOLGA2 - golgin A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004486.4 | 112 | Intron | NP_004477.3 | |||
XM_005251930.1 | 112 | Intron | XP_005251987.1 | |||
XM_005251931.1 | 112 | Intron | XP_005251988.1 | |||
XM_005251932.1 | 112 | Intron | XP_005251989.1 |
SWI5 - SWI5 homologous recombination repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040011.1 | 112 | Missense Mutation | GCG,TCG | A,S 38 | NP_001035100.1 | |
NM_001318089.1 | 112 | Missense Mutation | GCG,TCG | A,S 38 | NP_001305018.1 | |
NM_001318092.1 | 112 | Intron | NP_001305021.1 | |||
XM_011518652.2 | 112 | Missense Mutation | GCG,TCG | A,S 38 | XP_011516954.1 |