Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGTCAGTGTCCAAGCTACTGTC[A/G]GAGTGGCGCCCCAGTCAGGTTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609182 MIM: 610160 | ||||||||||||||||||||
Literature Links: |
SLC35D2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC35D2 - solute carrier family 35 member D2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF367 - zinc finger protein 367 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153695.3 | 1400 | Nonsense Mutation | CGA,TGA | R,* 349 | NP_710162.1 | |
XM_011518345.2 | 1400 | Intron | XP_011516647.1 |