Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGGTAGAAGAAATTACAGTTCAC[C/T]ACCTCCCTGTCACCTTTCCAGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603255 | ||||||||||||||||||||
Literature Links: |
NFX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NFX1 - nuclear transcription factor, X-box binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318758.1 | 308 | Missense Mutation | CCA,CTA | P,L 51 | NP_001305687.1 | |
NM_002504.5 | 308 | Missense Mutation | CCA,CTA | P,L 51 | NP_002495.2 | |
NM_147134.3 | 308 | Missense Mutation | CCA,CTA | P,L 51 | NP_667345.1 |