Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGAACCAGTTGCTACCTACTT[C/T]GCATACAGACTTGGTCCCCATGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604052 | ||||||||||||||||||||
Literature Links: |
TNFSF15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNFSF15 - tumor necrosis factor superfamily member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204344.1 | 723 | Missense Mutation | AAA,GAA | K,E 144 | NP_001191273.1 | |
NM_005118.3 | 723 | Missense Mutation | AAA,GAA | K,E 203 | NP_005109.2 |