Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTAGGATGAATTGCAGCTTAGA[A/G]GCATCAGGTATGGCAATGCGGTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607387 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID3C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID3C - AT-rich interaction domain 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DCTN3 - dynactin subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281425.1 | 302 | Silent Mutation | GCC,GCT | A,A 81 | NP_001268354.1 | |
NM_001281426.1 | 302 | Silent Mutation | GCC,GCT | A,A 81 | NP_001268355.1 | |
NM_001281427.1 | 302 | Silent Mutation | GCC,GCT | A,A 81 | NP_001268356.1 | |
NM_007234.4 | 302 | Silent Mutation | GCC,GCT | A,A 81 | NP_009165.1 | |
NM_024348.3 | 302 | Silent Mutation | GCC,GCT | A,A 81 | NP_077324.1 |
RPP25L - ribonuclease P/MRP subunit p25 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |