Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTGGGCTACCTCCTTGATGGTG[G/T]ACGCAGCTCTCTCCAGTTCTTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605712 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPTLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPTLC1 - serine palmitoyltransferase long chain base subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281303.1 | 1489 | Silent Mutation | GTA,GTC | V,V 451 | NP_001268232.1 | |
NM_006415.3 | 1489 | Missense Mutation | TAC,TCC | Y,S 462 | NP_006406.1 | |
NM_178324.2 | 1489 | Intron | NP_847894.1 | |||
XM_011518138.2 | 1489 | Intron | XP_011516440.1 | |||
XM_011518139.2 | 1489 | Missense Mutation | TAC,TCC | Y,S 307 | XP_011516441.1 | |
XM_017014200.1 | 1489 | Missense Mutation | TAC,TCC | Y,S 340 | XP_016869689.1 | |
XM_017014201.1 | 1489 | Missense Mutation | TAC,TCC | Y,S 340 | XP_016869690.1 | |
XM_017014202.1 | 1489 | Missense Mutation | TAC,TCC | Y,S 335 | XP_016869691.1 |