Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTACGAGGTCACCTGGGCTAACGA[C/T]GGCTACTGAGCACTCCCAGCCCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610167 MIM: 610615 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf172 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf172 - chromosome 9 open reading frame 172 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAMDC4 - MAM domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHPT1 - phosphohistidine phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135861.2 | 1039 | UTR 3 | NP_001129333.1 | |||
NM_001287342.1 | 1039 | Silent Mutation | GAC,GAT | D,D 125 | NP_001274271.1 | |
NM_001287343.1 | 1039 | UTR 3 | NP_001274272.1 | |||
NM_014172.5 | 1039 | Silent Mutation | GAC,GAT | D,D 123 | NP_054891.2 |
RABL6 - RAB, member RAS oncogene family-like 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |