Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCCTGTCGTCCCAGAGGTACT[C/T]GGGGGACAGCACCTTGGACGGCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606074 MIM: 601619 | ||||||||||||||||||||
Literature Links: |
GBGT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GBGT1 - globoside alpha-1,3-N-acetylgalactosaminyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282629.1 | 1298 | UTR 3 | NP_001269558.1 | |||
NM_001282632.1 | 1298 | Missense Mutation | NP_001269561.1 | |||
NM_001288572.1 | 1298 | Missense Mutation | NP_001275501.1 | |||
NM_001288573.1 | 1298 | Missense Mutation | NP_001275502.1 | |||
NM_021996.5 | 1298 | Missense Mutation | NP_068836.2 |
RALGDS - ral guanine nucleotide dissociation stimulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |