Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCTTGCTGTTCCTGCTCCTGCT[C/T]CCGCCGCTCCCCGTCCTGCTCGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176805 | ||||||||||||||||||||
Literature Links: |
PTGS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTGS1 - prostaglandin-endoperoxide synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000962.3 | 354 | Silent Mutation | CTC,CTT | L,L 16 | NP_000953.2 | |
NM_001271164.1 | 354 | Silent Mutation | CTC,CTT | L,L 16 | NP_001258093.1 | |
NM_001271165.1 | 354 | Intron | NP_001258094.1 | |||
NM_001271166.1 | 354 | UTR 5 | NP_001258095.1 | |||
NM_001271367.1 | 354 | UTR 5 | NP_001258296.1 | |||
NM_001271368.1 | 354 | UTR 5 | NP_001258297.1 | |||
NM_080591.2 | 354 | Silent Mutation | CTC,CTT | L,L 16 | NP_542158.1 | |
XM_005252105.3 | 354 | UTR 5 | XP_005252162.1 | |||
XM_011518875.2 | 354 | UTR 5 | XP_011517177.1 | |||
XM_011518876.2 | 354 | UTR 5 | XP_011517178.1 | |||
XM_017014929.1 | 354 | UTR 5 | XP_016870418.1 |