Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCATGGGGAACTGGATTCTGCTG[C/G]GAGCACAGGCTAAGGCTGTAGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604722 MIM: 608052 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D3C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SH2D3C - SH2 domain containing 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOR2A - torsin family 2 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085347.2 | 751 | Intron | NP_001078816.1 | |||
NM_001134430.2 | 751 | Intron | NP_001127902.1 | |||
NM_001134431.2 | 751 | UTR 3 | NP_001127903.1 | |||
NM_001252018.1 | 751 | Intron | NP_001238947.1 | |||
NM_001252021.1 | 751 | Intron | NP_001238950.1 | |||
NM_001252023.1 | 751 | Intron | NP_001238952.1 | |||
NM_130459.3 | 751 | Silent Mutation | TCC,TCG | S,S 224 | NP_569726.2 | |
XM_011518554.1 | 751 | Intron | XP_011516856.1 |
TTC16 - tetratricopeptide repeat domain 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |