Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGCTAGACCTCTTTCTCTAGCT[A/C]TATAACATTCACAAAATGACTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613574 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC39B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC39B - tetratricopeptide repeat domain 39B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168339.1 | Intron | NP_001161811.1 | ||||
NM_001168340.1 | Intron | NP_001161812.1 | ||||
NM_001168341.1 | Intron | NP_001161813.1 | ||||
NM_001168342.1 | Intron | NP_001161814.1 | ||||
NM_152574.2 | Intron | NP_689787.2 | ||||
XM_011517732.2 | Intron | XP_011516034.1 | ||||
XM_017014310.1 | Intron | XP_016869799.1 | ||||
XM_017014311.1 | Intron | XP_016869800.1 | ||||
XM_017014312.1 | Intron | XP_016869801.1 |