Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGAGGGTGTGGCACGTGGTCTG[C/G]AGCTGCTGAGTCAGGTTGCGGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLIN2 - perilipin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122.3 | 985 | Intron | NP_001113.2 | |||
XM_017014259.1 | 985 | Silent Mutation | CTC,CTG | L,L 321 | XP_016869748.1 |