Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAAGGACCCGCGGGCTTTTGGCT[A/G]GCAGCTGCTGCAGTGGCTCCTCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300371 MIM: 300398 | ||||||||||||||||||||
Literature Links: |
ABCD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCD1 - ATP binding cassette subfamily D member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000033.3 | 794 | Nonsense Mutation | TAG,TGG | *,W 132 | NP_000024.2 |
BCAP31 - B-cell receptor-associated protein 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139441.1 | 794 | Intron | NP_001132913.1 | |||
NM_001139457.2 | 794 | Intron | NP_001132929.1 | |||
NM_001256447.1 | 794 | Intron | NP_001243376.1 | |||
NM_005745.7 | 794 | Intron | NP_005736.3 |