Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATCCATAGACTGAACTTCTTCA[A/G]TTGAATCATCCAGGCTATTTGATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MAP7D2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP7D2 - MAP7 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168465.1 | 1934 | Missense Mutation | ACT,ATT | T,I 682 | NP_001161937.1 | |
NM_001168466.1 | 1934 | Missense Mutation | ACT,ATT | T,I 596 | NP_001161938.1 | |
NM_001168467.1 | 1934 | Missense Mutation | ACT,ATT | T,I 589 | NP_001161939.1 | |
NM_152780.3 | 1934 | Missense Mutation | ACT,ATT | T,I 641 | NP_689993.2 | |
XM_005274478.2 | 1934 | Missense Mutation | ACT,ATT | T,I 666 | XP_005274535.1 | |
XM_005274479.2 | 1934 | Missense Mutation | ACT,ATT | T,I 655 | XP_005274536.1 | |
XM_005274480.3 | 1934 | Missense Mutation | ACT,ATT | T,I 633 | XP_005274537.1 | |
XM_006724481.2 | 1934 | Missense Mutation | ACT,ATT | T,I 715 | XP_006724544.1 | |
XM_006724482.2 | 1934 | Missense Mutation | ACT,ATT | T,I 674 | XP_006724545.1 | |
XM_011545483.1 | 1934 | Missense Mutation | ACT,ATT | T,I 704 | XP_011543785.1 | |
XM_011545484.1 | 1934 | Missense Mutation | ACT,ATT | T,I 671 | XP_011543786.1 | |
XM_011545485.1 | 1934 | Missense Mutation | ACT,ATT | T,I 660 | XP_011543787.1 | |
XM_011545486.2 | 1934 | Intron | XP_011543788.1 | |||
XM_017029393.1 | 1934 | Missense Mutation | ACT,ATT | T,I 663 | XP_016884882.1 | |
XM_017029394.1 | 1934 | Missense Mutation | ACT,ATT | T,I 630 | XP_016884883.1 | |
XM_017029395.1 | 1934 | Missense Mutation | ACT,ATT | T,I 622 | XP_016884884.1 | |
XM_017029396.1 | 1934 | Missense Mutation | ACT,ATT | T,I 619 | XP_016884885.1 | |
XM_017029397.1 | 1934 | Missense Mutation | ACT,ATT | T,I 611 | XP_016884886.1 | |
XM_017029398.1 | 1934 | Intron | XP_016884887.1 |
MIR23C - microRNA 23c | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |