Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCTGCGGGCTGCGTGGCCGGGA[A/T]GAGCGCCAGCACGGGCGGTGGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300089 MIM: 300214 | ||||||||||||||||||||
Literature Links: |
IDH3G PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDH3G - isocitrate dehydrogenase 3 (NAD(+)) gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLXNB3 - plexin B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRPK3 - SRSF protein kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170760.1 | 88 | Missense Mutation | AAG,ATG | K,M 1 | NP_001164231.1 | |
NM_001170761.1 | 88 | Missense Mutation | AAG,ATG | K,M 1 | NP_001164232.1 | |
NM_014370.3 | 88 | Missense Mutation | AAG,ATG | K,M 1 | NP_055185.2 |