Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCAGGCCTTCGTGCACGTGGCC[A/G]TGGCCGGTGGCCTCTGTGCCGTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300059 | ||||||||||||||||||||
Literature Links: |
MIR3202-1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR3202-1 - microRNA 3202-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3202-2 - microRNA 3202-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM187 - transmembrane protein 187 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003492.2 | 589 | Missense Mutation | ATG,GTG | M,V 14 | NP_003483.1 | |
XM_011531199.2 | 589 | Missense Mutation | ATG,GTG | M,V 14 | XP_011529501.1 |