Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCACTCAATAACAGAGAGATCA[C/T]TGTGGTATTCTCTTCTCACGGCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300427 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLGN4X PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLGN4X - neuroligin 4, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282145.1 | Intron | NP_001269074.1 | ||||
NM_001282146.1 | Intron | NP_001269075.1 | ||||
NM_020742.3 | Intron | NP_065793.1 | ||||
NM_181332.2 | Intron | NP_851849.1 | ||||
XM_005274564.2 | Intron | XP_005274621.1 | ||||
XM_005274565.2 | Intron | XP_005274622.1 | ||||
XM_005274566.4 | Intron | XP_005274623.1 | ||||
XM_006724504.3 | Intron | XP_006724567.1 | ||||
XM_011545547.2 | Intron | XP_011543849.1 | ||||
XM_011545548.2 | Intron | XP_011543850.1 | ||||
XM_017029690.1 | Intron | XP_016885179.1 | ||||
XM_017029691.1 | Intron | XP_016885180.1 | ||||
XM_017029692.1 | Intron | XP_016885181.1 | ||||
XM_017029693.1 | Intron | XP_016885182.1 |