Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGGTCGCGCCATGGAGGGCGCCG[A/C]GCCCCGCGCGCGGCCCGAGCGCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300103 | ||||||||||||||||||||
Literature Links: |
SHROOM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHROOM2 - shroom family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320663.1 | 104 | Intron | NP_001307592.1 | |||
NM_001320664.1 | 104 | Intron | NP_001307593.1 | |||
NM_001649.3 | 104 | Missense Mutation | GAG,GCG | E,A 5 | NP_001640.1 | |
XM_005274500.4 | 104 | Missense Mutation | GAG,GCG | E,A 5 | XP_005274557.2 | |
XM_011545506.1 | 104 | Intron | XP_011543808.1 | |||
XM_017029492.1 | 104 | Intron | XP_016884981.1 | |||
XM_017029493.1 | 104 | Intron | XP_016884982.1 | |||
XM_017029494.1 | 104 | Intron | XP_016884983.1 |