Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGACAGGGACCATCTAAGCTATA[A/T]TCCACCAGAACAGCACACCATGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTCHD1-AS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTCHD1-AS - PTCHD1 antisense RNA (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF645 - zinc finger protein 645 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152577.3 | 618 | Missense Mutation | AAT,ATT | N,I 180 | NP_689790.1 |