Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTAAACACGAACCACAGGTTGC[G/C]GACCCAGAGCACCAGGTTCAGGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300111 MIM: 313475 | ||||||||||||||||||||
Literature Links: |
PRICKLE3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRICKLE3 - prickle planar cell polarity protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYP - synaptophysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003179.2 | 665 | Missense Mutation | CGC,GGC | R,G 217 | NP_003170.1 |
SYP-AS1 - SYP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |